Assay Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175121.3 | NP_780330.2 | 11 - 12 | 1323 | 73 | |
| GenBank mRNA | AK031615.1 | - | 11 - 12 | 1354 | 73 | |
| AK129342.1 | - | 11 - 12 | 1284 | 73 | ||
| AK164568.1 | - | 5 - 6 | 5086 | 73 | ||
| AK169378.1 | - | 1 - 2 | 82 | 73 | ||
| BC041108.1 | - | 11 - 12 | 1193 | 4038546 | 73 | |
| BC048178.1 | - | 11 - 12 | 1323 | 6825801 | 73 | |
| BC049271.1 | - | 4 - 5 | 346 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a2
Genomic Map