Assay Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175121.3 | NP_780330.2 | 5 - 6 | 766 | 69 | |
| GenBank mRNA | AK031615.1 | - | 5 - 6 | 797 | 69 | |
| AK129342.1 | - | 5 - 6 | 727 | 69 | ||
| BC037675.1 | - | 2 - 3 | 173 | 69 | ||
| BC041108.1 | - | 5 - 6 | 636 | 4038546 | 69 | |
| BC048178.1 | - | 5 - 6 | 766 | 6825801 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a2
Genomic Map