Assay Details
Gene Symbol: Med31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026068.2 | NP_080344.2 | 2 - 3 | 712 | 141 | |
GenBank mRNA | AK005655.1 | - | 2 - 3 | 121 | 141 | |
AK013990.1 | - | 2 - 3 | 153 | 141 | ||
BC019685.1 | - | 2 - 3 | 182 | 3983617 | 141 | |
BC051929.1 | - | 2 - 3 | 712 | 5709831 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910207 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910203 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910204 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910205 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910209 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910208 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910210 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910211 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910206 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | BC051929.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | NM_026068.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | AK005655.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | AK013990.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | BC019685.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | BC051929.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | NM_026068.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | AK005655.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | AK013990.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P255387 | BC019685.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773484 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 220481 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114422 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | BC051929.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | NM_026068.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | AK005655.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | AK013990.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | BC019685.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | BC051929.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | NM_026068.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | AK005655.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | AK013990.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P255387 | BC019685.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | BC051929.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | NM_026068.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | AK005655.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | AK013990.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | BC019685.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | BC051929.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | NM_026068.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | AK005655.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | AK013990.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P255387 | BC019685.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910206 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910202 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910190 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910198 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910204 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910228 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910199 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910214 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910234 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910201 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910217 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910209 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910221 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910227 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910231 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910226 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910194 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910237 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910223 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910213 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27910216 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114422 | - | - |
Whole Mouse Genome G4122A | A_51_P255387 | BC051929.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P255387 | NM_026068.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P255387 | AK005655.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P255387 | AK013990.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P255387 | BC019685.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P255387 | BC051929.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P255387 | NM_026068.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P255387 | AK005655.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P255387 | AK013990.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P255387 | BC019685.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139715 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136089 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114422 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09369798 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09369792 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09369807 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09369789 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09369791 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09369800 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09369804 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09369796 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09369799 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910203 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910208 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910207 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910204 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910209 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910206 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910205 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910211 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27910210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27910148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910238 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910194 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09369804 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09741474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09369799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910223 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09689391 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09011250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910187 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09741475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910204 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09606876 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09622441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09323319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910201 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910206 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27910241 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10388157 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10388159 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10388156 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10388158 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10378066 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10388155 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417387_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10378065 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10388154 | - | - |
Mouse Genome 430 2.0 Array | 1417387_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1417387_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67279_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67279_PM_at | - | - |
Mouse Expression Set 430 | 1417387_at | - | - |
Mouse Expression Set 430 | 1417387_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417387_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417387_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417387_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417387_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 921470 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6620014 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6620014 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006620014 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217567 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217567 | - | - |
mouse-6 v1.1 (Illumina) | scl39972.2.1_24-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6620014 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39972.2.1_24-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620014 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1217567 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217567 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl39972.2.1_24-S | - | - |
