Assay Details
| Gene Symbol: Adprhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133883.2 | NP_598644.1 | 5 - 6 | 851 | 103 | |
| GenBank mRNA | AJ427296.1 | - | 5 - 6 | 851 | 103 | |
| AK143583.1 | - | 5 - 6 | 841 | 103 | ||
| AK147034.1 | - | 5 - 6 | 827 | 103 | ||
| AK149904.1 | - | 3 - 4 | 1238 | 103 | ||
| AK150202.1 | - | 3 - 4 | 1238 | 103 | ||
| AK169070.1 | - | 5 - 6 | 838 | 103 | ||
| AK171780.1 | - | 5 - 6 | 841 | 103 | ||
| BC023177.1 | - | 5 - 6 | 828 | 4945959 | 103 | |
| BC028965.1 | - | 3 - 4 | 1010 | 103 | ||
| BC045203.1 | - | 5 - 6 | 889 | 5376002 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279471 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279470 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279469 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | BC028965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | BC023177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | BC045203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | AK150202.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | AK169070.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | BC028965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | BC023177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | BC045203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | AK150202.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520106 | AK169070.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874219 | BC120749.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874219 | BC120751.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874219 | NM_146550.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50528 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36903 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112883 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | BC028965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | BC023177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | BC045203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | AK150202.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | AK169070.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | BC028965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | BC023177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | BC045203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | AK150202.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520106 | AK169070.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | BC028965.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | BC023177.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | BC045203.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | AK150202.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | AK169070.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | BC028965.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | BC023177.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | BC045203.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | AK150202.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520106 | AK169070.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23279465 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112883 | - | - |
| Whole Mouse Genome G4122A | A_51_P520106 | BC028965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520106 | BC023177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520106 | BC045203.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520106 | AK150202.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520106 | AK169070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520106 | BC028965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520106 | BC023177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520106 | BC045203.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520106 | AK150202.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520106 | AK169070.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134954 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112883 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04026743 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03892079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04026755 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03892086 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03892082 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279469 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279470 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23279471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23279540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03892079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03892084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03916194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03892085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03842150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03972801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04026743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23279453 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10516333 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516334 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516329 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427449_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10516325 | - | - |
| Mouse Genome 430 2.0 Array | 1427449_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427449_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100206_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100206_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400028748.mm.1 | TC0400003535.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400028750.mm.1 | TC0400003535.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400028750.mm.1 | NM_133883.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028750.mm.1 | NM_133883.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028751.mm.1 | TC0400003535.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400028751.mm.1 | NM_133883.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028751.mm.1 | NM_133883.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028747.mm.1 | TC0400003535.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427449_a_at | - | - |
| Mouse Expression Set 430 | 1427449_PM_a_at | - | - |
| Murine Genome U74v2 Set | 102350_PM_at | - | - |
| Murine Genome U74v2 Set | 102349_at | - | - |
| Murine Genome U74v2 Set | 102349_PM_at | - | - |
| Murine Genome U74v2 Set | 102350_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427449_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427449_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427449_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427449_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 543152 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 130551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000130551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2639063 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2639063 | - | - |
| mouse-6 v1.1 (Illumina) | scl23822.4_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 130551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23822.4_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 130551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2639063 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2639063 | - | - |

Adprhl2
Genomic Map