Assay Details
Gene Symbol: Vmn1r76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134205.2 | NP_598966.2 | 1 | 443 | 108 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041205 | NM_134205.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180335 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041205 | NM_134205.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P162023 | NM_134205.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970144 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970152 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970136 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970147 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970145 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970132 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970134 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970126 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970135 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970138 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970140 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24970143 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180335 | - | - |
Whole Mouse Genome G4122A | A_51_P162023 | NM_134205.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180335 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06151911 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06605552 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112780 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06268818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24970152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970135 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05955343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05962378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05902976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970138 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970144 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06668053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06605549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06668058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06668054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06453636 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05955342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06499098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06668057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970136 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24970157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06036049 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10560014 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10560013 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700020791.mm.1 | TC0700002330.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700020791.mm.1 | NM_134205.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020791.mm.1 | NM_134205.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1230398 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1230398 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001230398 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252347 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1252347 | - | - |
mouse-6 v1.1 (Illumina) | scl9206.1.1_320-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1230398 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl9206.1.1_320-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1230398 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2460929 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2460929 | - | - |
