Assay Details
| Gene Symbol: Olfr803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146554.1 | NP_666765.1 | 1 | 535 | 139 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991457 | NM_146554.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155318 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991457 | NM_146554.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P415635 | NM_146554.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27546895 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155318 | - | - |
| Whole Mouse Genome G4122A | A_51_P415635 | NM_146554.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106947 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08265984 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08265983 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08265990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08265986 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08698836 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08265991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27546883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08907061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08265985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27546848 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373635 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373634 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 843815 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480279 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480279 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480279 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2718049 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2718049 | - | - |
| mouse-6 v1.1 (Illumina) | scl5159.1.1_233-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480279 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl5159.1.1_233-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480279 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2718049 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2718049 | - | - |

Olfr803
Genomic Map