Assay Details
| Gene Symbol: Olfr134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146832.1 | NP_667043.1 | 1 | 244 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P197735 | NM_146832.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P197735 | NM_146832.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P197735 | NM_146832.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P197735 | NM_146832.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318028 | NM_146832.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P197735 | NM_146832.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P197735 | NM_146832.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31171308 | - | - |
| Whole Mouse Genome G4122A | A_51_P318028 | NM_146832.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P197735 | NM_146832.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P197735 | NM_146832.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13380869 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13380875 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13371335 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13371326 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13380871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31171274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13481504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13297264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13470775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13297259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13552886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13242611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31171298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13497983 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445177 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445176 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445178 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700007779.mm.1 | NM_146832.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007779.mm.1 | NM_146832.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007779.mm.1 | TC1700000797.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 621431 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6350048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006350048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213093 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213093 | - | - |
| mouse-6 v1.1 (Illumina) | scl0258829.1_325-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6350048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258829.1_325-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350048 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213093 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213093 | - | - |

Olfr134
Genomic Map