Assay Details
| Gene Symbol: Olfr342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146948.1 | NP_667159.1 | 1 | 643 | 144 | |
| GenBank mRNA | BC120521.1 | - | 1 | 669 | 40129444 | 144 |
| BC120523.1 | - | 1 | 669 | 40129446 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P376207 | BC120521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P376207 | BC120523.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P376207 | NM_146948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P376207 | BC120521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P376207 | BC120523.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P376207 | NM_146948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P376207 | BC120521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P376207 | BC120523.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P376207 | NM_146948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P376207 | BC120521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P376207 | BC120523.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P376207 | NM_146948.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P376207 | BC120521.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P376207 | BC120523.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P376207 | NM_146948.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P376207 | BC120521.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P376207 | BC120523.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P376207 | NM_146948.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21139473 | - | - |
| Whole Mouse Genome G4122A | A_51_P376207 | BC120521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P376207 | BC120523.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P376207 | NM_146948.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P376207 | BC120521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P376207 | BC120523.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P376207 | NM_146948.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140457 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01567413 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01567387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01567396 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01567392 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01567388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21139489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01567390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01708716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01246128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01708714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02250551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21139489 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10471746 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10471745 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200006025.mm.1 | TC0200000661.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 405611 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5270692 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270692 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2741317 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2741317 | - | - |
| mouse-6 v1.1 (Illumina) | scl5052.1.1_79-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270692 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl5052.1.1_79-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270692 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2741317 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2741317 | - | - |

Olfr342
Genomic Map