| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700015033.mm.1 |
TC0700001766.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700015033.mm.1 |
NM_011247.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015033.mm.1 |
NM_011247.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015033.mm.1 |
NM_175023.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015033.mm.1 |
NM_175023.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015035.mm.1 |
TC0700001766.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700015035.mm.1 |
NM_011247.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015035.mm.1 |
NM_011247.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015035.mm.1 |
NM_175023.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015035.mm.1 |
NM_175023.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015034.mm.1 |
TC0700001766.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700015034.mm.1 |
NM_011247.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015034.mm.1 |
NM_011247.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015034.mm.1 |
NM_175023.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015034.mm.1 |
NM_175023.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015038.mm.1 |
TC0700001766.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700015038.mm.1 |
NM_011247.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015038.mm.1 |
NM_011247.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015038.mm.1 |
NM_175023.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015038.mm.1 |
NM_175023.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015036.mm.1 |
TC0700001766.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700015036.mm.1 |
NM_011247.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015036.mm.1 |
NM_011247.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015036.mm.1 |
NM_175023.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700015036.mm.1 |
NM_175023.3 |
Refseq |