Assay Details
Gene Symbol: Gas5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002840.2 | 3 - 4 | 2470 | 71 | ||
GenBank mRNA | AK083453.1 | - | 3 - 4 | 2471 | 71 | |
BC059726.1 | - | 7 - 8 | 259 | 71 | ||
BC080708.1 | - | 5 - 6 | 1527 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130544 | BC080708.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130544 | NR_002840.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130544 | AK083453.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130544 | BC080708.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130544 | NR_002840.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130544 | AK083453.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178378 | BC055883.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178378 | BC080708.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178378 | NR_002840.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178378 | AK083453.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134809 | BC055883.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134809 | BC059726.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134809 | BC080708.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134809 | NR_002840.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134809 | AK083453.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835520 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31178 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948064 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150025 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134809 | BC055883.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134809 | BC059726.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134809 | BC080708.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134809 | NR_002840.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134809 | AK083453.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178378 | BC055883.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178378 | BC080708.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178378 | NR_002840.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178378 | AK083453.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130544 | BC080708.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130544 | NR_002840.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130544 | AK083453.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130544 | BC080708.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130544 | NR_002840.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130544 | AK083453.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150025 | - | - |
Whole Mouse Genome G4122A | A_51_P366867 | - | - |
Whole Mouse Genome G4122A | A_51_P130544 | BC080708.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130544 | NR_002840.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130544 | AK083453.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130544 | BC080708.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130544 | NR_002840.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130544 | AK083453.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150025 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10351040 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351036 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351033 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351034 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351032 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351027 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351031 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351030 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351028 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351038 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351029 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424843_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449410_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419291_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436222_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10351037 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10351026 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10351035 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10351039 | - | - |
Mouse Genome 430 2.0 Array | 1419291_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1455904_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449410_a_at | - | - |
Mouse Genome 430 2.0 Array | 1449410_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1436222_at | - | - |
Mouse Genome 430 2.0 Array | 1419291_x_at | - | - |
Mouse Genome 430 2.0 Array | 1455904_at | - | - |
Mouse Genome 430 2.0 Array | 1436222_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1424843_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1424843_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14455_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14455_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100012042.mm.1 | NR_002840.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012042.mm.1 | NR_002840.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100012042.mm.1 | TC0100001464.mm.1 | MTA TC |
Mouse Expression Set 430 | 1449410_PM_a_at | - | - |
Mouse Expression Set 430 | 1419291_x_at | - | - |
Mouse Expression Set 430 | 1424843_a_at | - | - |
Mouse Expression Set 430 | 1455904_at | - | - |
Mouse Expression Set 430 | 1455904_PM_at | - | - |
Mouse Expression Set 430 | 1436222_at | - | - |
Mouse Expression Set 430 | 1449410_a_at | - | - |
Mouse Expression Set 430 | 1436222_PM_at | - | - |
Mouse Expression Set 430 | 1419291_PM_x_at | - | - |
Mouse Expression Set 430 | 1424843_PM_a_at | - | - |
Murine Genome U74v2 Set | 98531_g_at | - | - |
Murine Genome U74v2 Set | 101885_at | - | - |
Murine Genome U74v2 Set | 98530_PM_at | - | - |
Murine Genome U74v2 Set | 98530_at | - | - |
Murine Genome U74v2 Set | 101885_PM_at | - | - |
Murine Genome U74v2 Set | 98531_PM_g_at | - | - |
Mouse Genome 430A 2.0 Array | 1424843_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1449410_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1419291_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1436222_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1455904_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1436222_at | - | - |
Mouse Genome 430A 2.0 Array | 1455904_at | - | - |
Mouse Genome 430A 2.0 Array | 1419291_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1424843_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1449410_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424843_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1436222_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419291_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424843_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449410_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449410_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419291_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1436222_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 725392 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4210551 | - | - |
MouseWG-6 v2 Expression BeadChip | 1980136 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2680850 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2680850 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2680850 | - | - |
