Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400000613.mm.1 |
NM_001256142.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000613.mm.1 |
NM_001256142.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000613.mm.1 |
NM_001256145.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000613.mm.1 |
NM_001256145.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000613.mm.1 |
TC0400000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000610.mm.1 |
NM_001256142.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000610.mm.1 |
NM_001256142.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000610.mm.1 |
TC0400000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000612.mm.1 |
NM_001256142.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000612.mm.1 |
NM_001256142.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000612.mm.1 |
NM_001256145.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000612.mm.1 |
NM_001256145.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000612.mm.1 |
TC0400000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000614.mm.1 |
NM_001256142.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000614.mm.1 |
NM_001256142.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000614.mm.1 |
NM_001256145.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000614.mm.1 |
NM_001256145.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000614.mm.1 |
TC0400000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000611.mm.1 |
NM_001256142.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000611.mm.1 |
NM_001256142.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000611.mm.1 |
NM_001256145.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000611.mm.1 |
NM_001256145.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000611.mm.1 |
TC0400000091.mm.1 |
MTA TC |