Assay Details
Gene Symbol: Klk1b21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010642.2 | NP_034772.1 | 3 - 4 | 563 | 143 | |
GenBank mRNA | AB039276.1 | - | 3 - 4 | 566 | 143 | |
AK144221.1 | - | 3 - 4 | 530 | 143 | ||
BC012243.1 | - | 3 - 4 | 497 | 4167772 | 143 | |
BC145383.1 | - | 3 - 4 | 519 | 9053941 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136971 | NM_010642.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136971 | AB039276.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136971 | AK144221.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136971 | BC012243.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136971 | BC145383.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146772 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136971 | NM_010642.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136971 | AB039276.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136971 | AK144221.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136971 | BC012243.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136971 | BC145383.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085687 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085671 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085672 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085649 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146772 | - | - |
Whole Mouse Genome G4122A | A_52_P525574 | NM_010643.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P525574 | AB039277.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P525574 | BC146015.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P525574 | BC146019.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P525574 | NM_010642.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136971 | NM_010642.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136971 | AB039276.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136971 | AK144221.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136971 | BC012243.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136971 | BC145383.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085630 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06476303 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085691 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06350482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06204433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25085632 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10552593 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552588 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552591 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552589 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552592 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552590 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451607_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10552587 | - | - |
Mouse Genome 430 2.0 Array | 1451607_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1451607_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16616_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16616_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700006404.mm.1 | TC0700000789.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006404.mm.1 | NM_010642.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006403.mm.1 | TC0700000789.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006403.mm.1 | NM_010642.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006401.mm.1 | TC0700000789.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006401.mm.1 | NM_010642.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006405.mm.1 | TC0700000789.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006405.mm.1 | NM_010642.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006399.mm.1 | TC0700000789.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006399.mm.1 | NM_010642.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006402.mm.1 | TC0700000789.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006402.mm.1 | NM_010642.3 | Refseq |
Mouse Expression Set 430 | 1451607_PM_at | - | - |
Mouse Expression Set 430 | 1451607_at | - | - |
Murine Genome U74v2 Set | 162456_f_at | - | - |
Murine Genome U74v2 Set | 162456_PM_f_at | - | - |
Murine Genome U74v2 Set | 168876_PM_f_at | - | - |
Murine Genome U74v2 Set | 168876_f_at | - | - |
Murine Genome U74v2 Set | 170458_PM_r_at | - | - |
Murine Genome U74v2 Set | 170458_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1451607_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451607_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451607_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451607_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 528779 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 520632 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101740735 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1410541 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 520632 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000520632 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001410541 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2732087 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2651099 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1219551 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2651099 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2732087 | - | - |
mouse-6 v1.1 (Illumina) | GI_23956055-S | - | - |
mouse-6 v1.1 (Illumina) | scl0016616.1_34-S | - | - |
mouse-6 v1.1 (Illumina) | scl016616.4_12-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1410541 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 520632 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101740735 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl016616.4_12-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0016616.1_34-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101740735 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1410541 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520632 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2651099 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2732087 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2732087 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219551 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2651099 | - | - |
