| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700005951.mm.1 |
TC0700000733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005951.mm.1 |
NM_023363.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005951.mm.1 |
NM_023363.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005954.mm.1 |
TC0700000733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005954.mm.1 |
NM_023363.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005954.mm.1 |
NM_023363.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005946.mm.1 |
TC0700000733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005946.mm.1 |
NM_023363.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005946.mm.1 |
NM_023363.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005952.mm.1 |
TC0700000733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005952.mm.1 |
NM_023363.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005952.mm.1 |
NM_023363.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005948.mm.1 |
TC0700000733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005948.mm.1 |
NM_023363.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005948.mm.1 |
NM_023363.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005941.mm.1 |
TC0700000733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005941.mm.1 |
NM_023363.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005941.mm.1 |
NM_023363.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005945.mm.1 |
TC0700000733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005945.mm.1 |
NM_023363.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005945.mm.1 |
NM_023363.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005953.mm.1 |
TC0700000733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005953.mm.1 |
NM_023363.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005953.mm.1 |
NM_023363.2 |
Refseq |