Assay Details
| Gene Symbol: Zfp788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023363.2 | NP_075852.1 | 5 - 6 | 481 | 120 | |
| GenBank mRNA | AK020228.1 | - | 4 - 5 | 352 | 120 | |
| AK036346.1 | - | 3 - 4 | 310 | 120 | ||
| AK082981.1 | - | 5 - 6 | 482 | 120 | ||
| AK163311.1 | - | 5 - 6 | 428 | 120 | ||
| BC027798.1 | - | 2 - 3 | 210 | 5323247 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071863 | BC027798.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071863 | AK036346.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071863 | NM_023363.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689242 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116379 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071863 | BC027798.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071863 | AK036346.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071863 | NM_023363.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25078043 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06175377 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161353 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154766 | - | - |
| Whole Mouse Genome G4122A | A_51_P453935 | BC027798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P453935 | AK082981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P453935 | AK163311.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P453935 | AK036346.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P453935 | NM_023363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P361066 | BC027798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P361066 | AK082981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P361066 | AK163311.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P361066 | AK036346.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P361066 | NM_023363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P409906 | BC027798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P409906 | AK082981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P409906 | AK163311.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P409906 | AK036346.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P409906 | NM_023363.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124766 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154766 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06486792 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06486803 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06486791 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06486794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25077997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06482209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06336096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06305851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06486792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06486791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06180626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06209454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078032 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10536078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552325 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536075 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536077 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552321 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429371_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441434_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10536068 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552320 | - | - |
| Mouse Genome 430 2.0 Array | 1441434_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429371_at | - | - |
| Mouse Genome 430 2.0 Array | 1429371_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441434_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67607_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67607_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700005956.mm.1 | TC0700000733.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005956.mm.1 | NM_023363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005956.mm.1 | NM_023363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005958.mm.1 | TC0700000733.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005958.mm.1 | NM_023363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005958.mm.1 | NM_023363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005961.mm.1 | TC0700000733.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005961.mm.1 | NM_023363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005961.mm.1 | NM_023363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005957.mm.1 | TC0700000733.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005957.mm.1 | NM_023363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005957.mm.1 | NM_023363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005960.mm.1 | TC0700000733.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005960.mm.1 | NM_023363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005960.mm.1 | NM_023363.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005955.mm.1 | TC0700000733.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700005955.mm.1 | NM_023363.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005955.mm.1 | NM_023363.2 | Refseq |
| Mouse Expression Set 430 | 1441434_at | - | - |
| Mouse Expression Set 430 | 1429371_at | - | - |
| Mouse Expression Set 430 | 1441434_PM_at | - | - |
| Mouse Expression Set 430 | 1429371_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441434_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429371_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441434_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429371_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 366561 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1990605 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5860019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001990605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005860019 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2656296 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2626713 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2626713 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2656296 | - | - |
| mouse-6 v1.1 (Illumina) | scl000270.1_3-S | - | - |
| mouse-6 v1.1 (Illumina) | scl32762.6_595-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5860019 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1990605 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32762.6_595-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000270.1_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990605 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2626713 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2656296 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2626713 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2656296 | - | - |

Zfp788
Genomic Map