Assay Details
| Gene Symbol: Usp47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133758.3 | NP_598519.2 | 20 - 21 | 3298 | 81 | |
| NM_177249.3 | NP_796223.2 | 21 - 22 | 3358 | 81 | ||
| GenBank mRNA | AK030909.1 | - | 5 - 6 | 1249 | 81 | |
| AK030963.1 | - | 1 - 2 | 130 | 81 | ||
| AK131138.1 | - | 11 - 12 | 1975 | 81 | ||
| BC024117.1 | - | 4 - 5 | 1124 | 81 | ||
| BC057362.1 | - | 1 - 2 | 382 | 81 | ||
| BC089364.1 | - | 1 - 2 | 172 | 81 | ||
| BC108425.1 | - | 21 - 22 | 3299 | 30062566 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp47
Genomic Map