Assay Details
Gene Symbol: Fam134a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170755.2 | NP_739561.2 | 8 - 9 | 1073 | 85 | |
GenBank mRNA | AK034667.1 | - | 8 - 9 | 1075 | 86 | |
AK158940.1 | - | 8 - 9 | 842 | 85 | ||
AK160400.1 | - | 8 - 9 | 1027 | 85 | ||
AK165957.1 | - | 8 - 9 | 1052 | 85 | ||
AK172093.1 | - | 8 - 9 | 1070 | 85 | ||
AK191491.1 | - | 1 - 2 | 49 | 85 | ||
AK210282.1 | - | 1 - 2 | 49 | 85 | ||
AK210397.1 | - | 1 - 2 | 49 | 85 | ||
AK218481.1 | - | 1 - 2 | 49 | 85 | ||
BC026519.1 | - | 4 - 5 | 424 | 85 | ||
BC038286.1 | - | 4 - 5 | 459 | 4221441 | 85 | |
BC070406.1 | - | 8 - 9 | 1065 | 5707319 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352249 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352250 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352248 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352251 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | BC026519.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | BC038286.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | BC070406.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | AK172093.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | AK160400.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | BC026519.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | BC038286.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | BC070406.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | AK172093.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174415 | AK160400.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986466 | BC026519.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986466 | BC038286.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986466 | BC070406.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986466 | AK172093.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986466 | AK160400.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256376 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634465 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139241 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986466 | BC026519.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986466 | BC038286.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986466 | BC070406.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986466 | AK172093.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986466 | AK160400.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | BC026519.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | BC038286.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | BC070406.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | AK172093.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | AK160400.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | BC026519.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | BC038286.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | BC070406.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | AK172093.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174415 | AK160400.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352285 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352280 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352278 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352277 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352279 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139241 | - | - |
Whole Mouse Genome G4122A | A_51_P174415 | BC026519.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P174415 | BC038286.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P174415 | BC070406.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P174415 | AK172093.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P174415 | AK160400.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P174415 | BC026519.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P174415 | BC038286.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P174415 | BC070406.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P174415 | AK172093.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P174415 | AK160400.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103363 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139241 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00789304 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00789308 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352249 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352250 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352251 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20352248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00772945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20352252 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20352288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01081245 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00810533 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01081239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00706034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20352296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00574190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00810535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00122530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01081240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00122527 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00574189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00574186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20352279 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00706028 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00789304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20352297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00122529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01081241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00122528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20352300 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01081243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20352293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00904706 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20352282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00923527 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01081242 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00706029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20352290 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00706032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00810541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00923528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00574187 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10347514 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347515 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347510 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347518 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347509 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347517 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347512 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347513 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347519 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347511 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347516 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10347520 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452164_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10347508 | - | - |
Mouse Genome 430 2.0 Array | 1452164_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452164_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 227298_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 227298_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100005387.mm.1 | NM_170755.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100005387.mm.1 | NM_170755.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100005387.mm.1 | TC0100000652.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100005386.mm.1 | NM_170755.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100005386.mm.1 | NM_170755.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100005386.mm.1 | TC0100000652.mm.1 | MTA TC |
Mouse Expression Set 430 | 1452164_PM_at | - | - |
Mouse Expression Set 430 | 1452164_at | - | - |
Murine Genome U74v2 Set | 95401_at | - | - |
Murine Genome U74v2 Set | 95401_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452164_at | - | - |
Mouse Genome 430A 2.0 Array | 1452164_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452164_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452164_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3290673 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3290673 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003290673 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213426 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1213426 | - | - |
mouse-6 v1.1 (Illumina) | scl17783.10_25-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3290673 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl17783.10_25-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290673 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1213426 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213426 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl17783.10_25-S | - | - |
