Assay Details
| Gene Symbol: Hmbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110251.1 | NP_001103721.1 | 4 - 5 | 330 | 69 | |
| NM_013551.2 | NP_038579.2 | 4 - 5 | 388 | 69 | ||
| GenBank mRNA | AK028234.1 | - | 4 - 5 | 255 | 69 | |
| AK143558.1 | - | 4 - 5 | 255 | 69 | ||
| AK151020.1 | - | 4 - 5 | 354 | 69 | ||
| AK153884.1 | - | 4 - 5 | 320 | 69 | ||
| AK166734.1 | - | 4 - 5 | 286 | 69 | ||
| AK167702.1 | - | 4 - 5 | 347 | 69 | ||
| BB844509.1 | - | 4 - 5 | 331 | 69 | ||
| BC003861.1 | - | 4 - 5 | 348 | 3487642 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmbs
Genomic Map