Assay Details
| Gene Symbol: Prkar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021880.2 | NP_068680.1 | 6 - 7 | 680 | 106 | |
| GenBank mRNA | AK005039.1 | - | 6 - 7 | 682 | 106 | |
| AK019147.1 | - | 4 - 5 | 315 | 106 | ||
| AK027916.1 | - | 6 - 7 | 647 | 106 | ||
| AK049832.1 | - | 6 - 7 | 673 | 106 | ||
| AK051068.1 | - | 6 - 7 | 681 | 106 | ||
| AK053918.1 | - | 6 - 7 | 681 | 106 | ||
| AK083383.1 | - | 6 - 7 | 661 | 106 | ||
| AK145797.1 | - | 6 - 7 | 681 | 106 | ||
| AK145860.1 | - | 6 - 7 | 648 | 106 | ||
| AK146380.1 | - | 6 - 7 | 667 | 106 | ||
| AK147188.1 | - | 6 - 7 | 672 | 106 | ||
| AK150427.1 | - | 6 - 7 | 673 | 106 | ||
| AK150927.1 | - | 4 - 5 | 313 | 106 | ||
| AK151040.1 | - | 4 - 5 | 313 | 106 | ||
| AK151124.1 | - | 6 - 7 | 639 | 106 | ||
| AK151556.1 | - | 4 - 5 | 313 | 106 | ||
| AK152044.1 | - | 4 - 5 | 313 | 106 | ||
| AK153227.1 | - | 6 - 7 | 639 | 106 | ||
| AK158562.1 | - | 6 - 7 | 1018 | 106 | ||
| BC003461.1 | - | 6 - 7 | 651 | 2648357 | 106 | |
| BC005697.1 | - | 6 - 7 | 622 | 3597765 | 106 | |
| HQ412666.1 | - | 7 - 8 | 863 | 106 | ||
| HQ412667.1 | - | 6 - 7 | 679 | 106 | ||
| HQ412668.1 | - | 6 - 7 | 739 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkar1a
Genomic Map