Assay Details
| Gene Symbol: Mrps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025305.2 | NP_079581.1 | 4 - 5 | 514 | 89 | |
| GenBank mRNA | AK009251.1 | - | 4 - 5 | 479 | 89 | |
| AK012225.1 | - | 4 - 5 | 514 | 89 | ||
| AK131937.1 | - | 4 - 5 | 525 | 89 | ||
| AK162024.1 | - | 4 - 5 | 493 | 89 | ||
| BC049636.1 | - | 4 - 5 | 514 | 6532964 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154365 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154362 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978236 | AK012225.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978236 | AK162024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978236 | BC049636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978236 | NM_025305.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978236 | AK009251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193302 | AK162024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193302 | BC049636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193302 | NM_025305.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193302 | AK009251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193302 | AK162024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193302 | BC049636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193302 | NM_025305.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193302 | AK009251.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641256 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108909 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978236 | AK012225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978236 | AK162024.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978236 | BC049636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978236 | NM_025305.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978236 | AK009251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193302 | AK162024.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193302 | BC049636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193302 | NM_025305.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193302 | AK009251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193302 | AK162024.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193302 | BC049636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193302 | NM_025305.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193302 | AK009251.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P26859 | AK012225.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P26859 | AK131937.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P26859 | AK162024.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P26859 | BC049636.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P26859 | NM_025305.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193302 | AK162024.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193302 | BC049636.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193302 | NM_025305.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193302 | AK009251.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193302 | AK162024.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193302 | BC049636.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193302 | NM_025305.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193302 | AK009251.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28154374 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108909 | - | - |
| Whole Mouse Genome G4122A | A_52_P26859 | AK012225.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P26859 | AK131937.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P26859 | AK162024.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P26859 | BC049636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P26859 | NM_025305.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193302 | AK162024.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193302 | BC049636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193302 | NM_025305.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193302 | AK009251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193302 | AK162024.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193302 | BC049636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193302 | NM_025305.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193302 | AK009251.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116414 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108909 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09406571 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09406569 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09406568 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154365 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28154362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28154410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09223049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09271815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09406572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09406566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28154376 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10382570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382569 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453725_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10382565 | - | - |
| Mouse Genome 430 2.0 Array | 1453725_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1453725_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50529_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50529_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100017680.mm.1 | NM_025305.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017680.mm.1 | TC1100004280.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100017679.mm.1 | NM_025305.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017679.mm.1 | TC1100004280.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453725_PM_a_at | - | - |
| Mouse Expression Set 430 | 1453725_a_at | - | - |
| Murine Genome U74v2 Set | 96904_at | - | - |
| Murine Genome U74v2 Set | 96904_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453725_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453725_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453725_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453725_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 379870 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4730270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4730270 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004730270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2686710 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2686710 | - | - |
| mouse-6 v1.1 (Illumina) | scl40724.4.1_15-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4730270 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40724.4.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4730270 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2981821 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2981818 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2981818 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2981821 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40724.4.1_15-S | - | - |

Mrps7
Genomic Map