Assay Details
| Gene Symbol: Prpf38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025845.2 | NP_080121.1 | 1 - 2 | 547 | 71 | |
| GenBank mRNA | AK015938.1 | - | 1 - 2 | 554 | 71 | |
| AK018126.1 | - | 1 - 2 | 571 | 71 | ||
| AK159267.1 | - | 1 - 2 | 570 | 71 | ||
| BC050900.1 | - | 1 - 2 | 547 | 6402417 | 71 | |
| BC052077.1 | - | 1 - 2 | 547 | 6402417 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf38b
Genomic Map