Assay Details
| Gene Symbol: Tbc1d23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026254.2 | NP_080530.2 | 2 - 3 | 258 | 111 | |
| GenBank mRNA | AK155142.1 | - | 2 - 3 | 279 | 111 | |
| AK171591.1 | - | 2 - 3 | 166 | 111 | ||
| BC031706.1 | - | 2 - 3 | 258 | 4194266 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d23
Genomic Map