Assay Details
| Gene Symbol: Srrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026886.3 | NP_081162.1 | 7 - 8 | 959 | 88 | |
| GenBank mRNA | AK053836.1 | - | 7 - 8 | 691 | 88 | |
| AK165719.1 | - | 7 - 8 | 677 | 88 | ||
| BC056392.1 | - | 7 - 8 | 949 | 6855144 | 88 | |
| JN860221.1 | - | 7 - 8 | 606 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srrm4
Genomic Map