Assay Details
| Gene Symbol: Srrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026886.3 | NP_081162.1 | 9 - 10 | 1422 | 129 | |
| GenBank mRNA | AK053836.1 | - | 9 - 10 | 1154 | 129 | |
| AK129457.1 | - | 1 - 2 | 504 | 129 | ||
| BC056392.1 | - | 9 - 10 | 1412 | 6855144 | 129 | |
| JN860221.1 | - | 9 - 10 | 1069 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srrm4
Genomic Map