Assay Details
| Gene Symbol: Snx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026912.1 | NP_081188.1 | 2 - 3 | 206 | 71 | |
| GenBank mRNA | BC016091.1 | - | 2 - 3 | 206 | 4503873 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Snx15
Genomic Map