Assay Details
| Gene Symbol: Dhx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026987.2 | NP_081263.2 | 13 - 14 | 2287 | 67 | |
| GenBank mRNA | AK122314.2 | - | 13 - 14 | 2222 | 67 | |
| BC009147.1 | - | 13 - 14 | 2246 | 2647184 | 67 | |
| BC046781.1 | - | 13 - 14 | 2215 | 5716182 | 67 | |
| BC138204.1 | - | 13 - 14 | 2226 | 8861223 | 67 | |
| BC138205.1 | - | 13 - 14 | 2226 | 8861224 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dhx16
Genomic Map