Assay Details
| Gene Symbol: Dhx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026987.2 | NP_081263.2 | 7 - 8 | 1459 | 66 | |
| GenBank mRNA | AK122314.2 | - | 7 - 8 | 1422 | 66 | |
| AK178678.1 | - | 1 - 2 | 87 | 66 | ||
| BC009147.1 | - | 7 - 8 | 1418 | 2647184 | 66 | |
| BC046781.1 | - | 7 - 8 | 1412 | 5716182 | 66 | |
| BC138204.1 | - | 7 - 8 | 1398 | 8861223 | 66 | |
| BC138205.1 | - | 7 - 8 | 1398 | 8861224 | 66 | |
| BU054791.1 | - | 2 - 3 | 292 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhx16
Genomic Map