Assay Details
| Gene Symbol: Ppme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028292.2 | NP_082568.1 | 1 - 2 | 173 | 100 | |
| GenBank mRNA | AK012256.1 | - | 1 - 2 | 245 | 100 | |
| AK076992.1 | - | 1 - 2 | 584 | 100 | ||
| AK146051.1 | - | 1 - 2 | 155 | 100 | ||
| AK146268.1 | - | 1 - 2 | 185 | 100 | ||
| AK154876.1 | - | 1 - 2 | 246 | 100 | ||
| BC014867.1 | - | 1 - 2 | 173 | 3596696 | 100 | |
| BC029064.1 | - | 1 - 2 | 161 | 4952806 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppme1
Genomic Map