Assay Details
| Gene Symbol: Pdia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028295.1 | NP_082571.1 | 2 - 3 | 276 | 67 | |
| GenBank mRNA | BC009151.1 | - | 2 - 3 | 276 | 3600849 | 67 |
More Information
Set Membership: |
|

Pdia5
Genomic Map