Assay Details
| Gene Symbol: Ccdc181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029115.3 | NP_083391.2 | 2 - 3 | 307 | 125 | |
| GenBank mRNA | AK015464.1 | - | 2 - 3 | 294 | 125 | |
| AK076640.1 | - | 2 - 3 | 302 | 125 | ||
| BC048086.1 | - | 2 - 3 | 199 | 6390120 | 125 | |
| BC060044.1 | - | 2 - 3 | 306 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P190437 | BC060044.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P190437 | AK076640.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P190437 | BC048086.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P190437 | NM_029115.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P190437 | BC060044.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P190437 | AK076640.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P190437 | BC048086.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P190437 | NM_029115.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494799 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27364 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118740 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100673 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P190437 | BC060044.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P190437 | AK076640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P190437 | BC048086.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P190437 | NM_029115.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P190437 | BC060044.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P190437 | AK076640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P190437 | BC048086.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P190437 | NM_029115.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20788170 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04337598 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100673 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118740 | - | - |
| Whole Mouse Genome G4122A | A_51_P190437 | BC060044.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190437 | AK076640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190437 | BC048086.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190437 | NM_029115.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190437 | BC060044.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190437 | AK076640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190437 | BC048086.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190437 | NM_029115.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101392 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117539 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100673 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118740 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00219380 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00219382 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00219379 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00219386 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00075343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20788338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00219384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00223751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00333743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00922131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00922133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00461291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01124674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01091569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00922143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20788198 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351272 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351275 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351276 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351274 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351270 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351273 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435052_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351269 | - | - |
| Mouse Genome 430 2.0 Array | 1435052_at | - | - |
| Mouse Genome 430 2.0 Array | 1435052_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74895_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74895_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100012380.mm.1 | NM_029115.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012380.mm.1 | NM_029115.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012380.mm.1 | TC0100001501.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435052_PM_at | - | - |
| Mouse Expression Set 430 | 1435052_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435052_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435052_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 328824 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1450446 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450446 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769153 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2769153 | - | - |
| mouse-6 v1.1 (Illumina) | scl17282.7.92_117-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450446 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17282.7.92_117-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450446 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2908953 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2769153 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2908953 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl17282.7.92_117-S | - | - |

Ccdc181
Genomic Map