| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500012696.mm.1 |
NM_001164641.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012696.mm.1 |
NM_001164641.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012696.mm.1 |
NM_029640.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012696.mm.1 |
NM_029640.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012696.mm.1 |
TC1500001684.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_001164641.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_001164641.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_001164642.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_001164642.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_001164643.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_001164643.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_029640.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_029640.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_180662.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
NM_180662.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012697.mm.1 |
TC1500001684.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012699.mm.1 |
NM_029640.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012699.mm.1 |
NM_029640.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012699.mm.1 |
TC1500001684.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_001164641.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_001164641.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_001164642.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_001164642.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_001164643.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_001164643.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_029640.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_029640.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_180662.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
NM_180662.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012695.mm.1 |
TC1500001684.mm.1 |
MTA TC |