Assay Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282126.1 | NP_001269055.1 | 12 - 13 | 4002 | 80 | |
| NM_001282127.1 | NP_001269056.1 | 13 - 14 | 3665 | 80 | ||
| NM_001282128.1 | NP_001269057.1 | 13 - 14 | 3915 | 80 | ||
| NM_030178.2 | NP_084454.1 | 12 - 13 | 3767 | 80 | ||
| GenBank mRNA | AK019522.1 | - | 7 - 8 | 1292 | 80 | |
| AK047173.1 | - | 1 - 2 | 2027 | 80 | ||
| AK139006.1 | - | 1 - 2 | 1497 | 80 | ||
| AK156375.1 | - | 12 - 13 | 3950 | 80 | ||
| BC005647.1 | - | 11 - 12 | 1937 | 80 | ||
| BC031594.1 | - | 10 - 11 | 2691 | 80 | ||
| BC046521.1 | - | 12 - 13 | 3707 | 5363697 | 80 | |
| BC051640.1 | - | 1 - 2 | 101 | 80 | ||
| BC138361.1 | - | 13 - 14 | 3489 | 8861381 | 80 | |
| BC145256.1 | - | 13 - 14 | 3739 | 9053809 | 80 | |
| DQ288860.1 | - | 8 - 9 | 2355 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116853 | BC005647.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116853 | BC031594.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116853 | BC046521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116853 | AK047173.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116853 | AK139006.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371341 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489384 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619873 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624321 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234406 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59658 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156912 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140927 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111698 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156299 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116853 | BC005647.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116853 | BC031594.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116853 | BC046521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116853 | AK047173.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116853 | AK139006.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271354 | BC005647.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271354 | BC031594.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271354 | BC046521.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271354 | BC138361.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271354 | BC145256.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758791 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06030617 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140927 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111698 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156299 | - | - |
| Whole Mouse Genome G4122A | A_51_P271354 | BC005647.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271354 | BC031594.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271354 | BC046521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271354 | BC138361.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271354 | BC145256.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140028 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136878 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156299 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140927 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05500218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05500225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05122274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05144422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05500217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05131640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758761 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10540623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540631 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540630 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540629 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540638 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460402_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10540622 | - | - |
| Mouse Genome 430 2.0 Array | 1460402_at | - | - |
| Mouse Genome 430 2.0 Array | 1460402_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78783_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78783_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600009628.mm.1 | TC0600001245.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600009628.mm.1 | NM_001282126.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009628.mm.1 | NM_001282127.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009628.mm.1 | NM_001282128.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009628.mm.1 | NM_030178.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009629.mm.1 | TC0600001245.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600009629.mm.1 | NM_001282126.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009629.mm.1 | NM_001282127.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009629.mm.1 | NM_001282128.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009629.mm.1 | NM_030178.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009633.mm.1 | TC0600001245.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600009633.mm.1 | NM_001282126.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009633.mm.1 | NM_001282127.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009633.mm.1 | NM_001282128.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009633.mm.1 | NM_030178.2 | Refseq |
| Mouse Expression Set 430 | 1460402_PM_at | - | - |
| Mouse Expression Set 430 | 1460402_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460402_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460402_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460402_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460402_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3170139 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3170139 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170139 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236149 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236149 | - | - |
| mouse-6 v1.1 (Illumina) | scl29635.14.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170139 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29635.14.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170139 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2800859 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2800861 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2800859 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2800861 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29635.14.1_23-S | - | - |

Brpf1
Genomic Map