Assay Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282126.1 | NP_001269055.1 | 4 - 5 | 2161 | 75 | |
| NM_001282127.1 | NP_001269056.1 | 4 - 5 | 1929 | 75 | ||
| NM_001282128.1 | NP_001269057.1 | 4 - 5 | 2161 | 75 | ||
| NM_030178.2 | NP_084454.1 | 4 - 5 | 1929 | 75 | ||
| GenBank mRNA | AK043372.1 | - | 4 - 5 | 2162 | 75 | |
| AK156375.1 | - | 4 - 5 | 2109 | 75 | ||
| BC005647.1 | - | 2 - 3 | 201 | 75 | ||
| BC031594.1 | - | 2 - 3 | 835 | 75 | ||
| BC046521.1 | - | 4 - 5 | 1869 | 5363697 | 75 | |
| BC138361.1 | - | 4 - 5 | 1753 | 8861381 | 75 | |
| BC145256.1 | - | 4 - 5 | 1985 | 9053809 | 75 | |
More Information
Set Membership: |
|

Brpf1
Genomic Map