Assay Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271873.1 | NP_001258802.1 | 4 - 5 | 606 | 82 | |
| NM_133979.3 | NP_598740.1 | 4 - 5 | 606 | 82 | ||
| GenBank mRNA | AK043011.1 | - | 4 - 5 | 569 | 82 | |
| AK046091.1 | - | 4 - 5 | 558 | 82 | ||
| AK079622.1 | - | 4 - 5 | 557 | 82 | ||
| AK080947.1 | - | 4 - 5 | 558 | 82 | ||
| BC099688.1 | - | 4 - 5 | 529 | 6313494 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ano10
Genomic Map