Assay Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271873.1 | NP_001258802.1 | 6 - 7 | 1119 | 88 | |
| NM_133979.3 | NP_598740.1 | 6 - 7 | 1293 | 88 | ||
| GenBank mRNA | AK028391.1 | - | 1 - 2 | 238 | 88 | |
| AK043011.1 | - | 6 - 7 | 1256 | 88 | ||
| AK046091.1 | - | 6 - 7 | 1245 | 88 | ||
| AK054163.1 | - | 2 - 3 | 583 | 88 | ||
| AK079622.1 | - | 6 - 7 | 1244 | 88 | ||
| AK080947.1 | - | 6 - 7 | 1245 | 88 | ||
| AK138414.1 | - | 6 - 7 | 1368 | 88 | ||
| AK192365.1 | - | 1 - 2 | 37 | 88 | ||
| BC026421.1 | - | 1 - 2 | 333 | 88 | ||
| BC099688.1 | - | 6 - 7 | 1042 | 6313494 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ano10
Genomic Map