Assay Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271873.1 | NP_001258802.1 | 7 - 8 | 1176 | 69 | |
| NM_133979.3 | NP_598740.1 | 7 - 8 | 1350 | 69 | ||
| GenBank mRNA | AK028391.1 | - | 2 - 3 | 295 | 69 | |
| AK043011.1 | - | 7 - 8 | 1313 | 69 | ||
| AK046091.1 | - | 7 - 8 | 1302 | 69 | ||
| AK054163.1 | - | 3 - 4 | 640 | 69 | ||
| AK079622.1 | - | 7 - 8 | 1301 | 69 | ||
| AK080947.1 | - | 7 - 8 | 1302 | 69 | ||
| AK138414.1 | - | 7 - 8 | 1425 | 69 | ||
| BC026421.1 | - | 2 - 3 | 390 | 69 | ||
| BC099688.1 | - | 7 - 8 | 1099 | 6313494 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ano10
Genomic Map