Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 9 - 10 | 1479 | 85 | |
| NM_138306.2 | NP_612179.2 | 8 - 9 | 878 | 85 | ||
| GenBank mRNA | AK087503.1 | - | 9 - 10 | 1480 | 85 | |
| AK170936.1 | - | 5 - 6 | 501 | 85 | ||
| BC049228.1 | - | 8 - 9 | 914 | 3668704 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map