Assay Details
| Gene Symbol: Kif22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145588.1 | NP_663563.1 | 10 - 11 | 1620 | 73 | |
| GenBank mRNA | AK075922.1 | - | 10 - 11 | 1611 | 73 | |
| AK088033.1 | - | 7 - 8 | 1196 | 73 | ||
| AK205411.1 | - | 1 - 2 | 70 | 73 | ||
| BC003427.1 | - | 10 - 11 | 1620 | 2615715 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kif22
Genomic Map