Assay Details
| Gene Symbol: Kif22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145588.1 | NP_663563.1 | 11 - 12 | 1687 | 63 | |
| GenBank mRNA | AK075922.1 | - | 11 - 12 | 1678 | 63 | |
| AK084310.1 | - | 11 - 12 | 1676 | 63 | ||
| AK088033.1 | - | 8 - 9 | 1263 | 63 | ||
| BC003427.1 | - | 11 - 12 | 1687 | 2615715 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kif22
Genomic Map