Assay Details
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027905.2 | NP_082181.2 | 1 - 2 | 555 | 125 | |
GenBank mRNA | AK005048.1 | - | 1 - 2 | 555 | 125 | |
AK054432.1 | - | 1 - 2 | 575 | 125 | ||
AK088096.1 | - | 1 - 2 | 580 | 125 | ||
AK161059.1 | - | 1 - 2 | 564 | 125 | ||
BC141293.1 | - | 1 - 2 | 624 | 9055905 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
