Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500006776.mm.1 |
NM_027905.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006776.mm.1 |
TC1500000862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500006777.mm.1 |
NM_027905.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006777.mm.1 |
TC1500000862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500006778.mm.1 |
NM_027905.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006778.mm.1 |
TC1500000862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500006774.mm.1 |
NM_027905.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006774.mm.1 |
TC1500000862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500006775.mm.1 |
NM_027905.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006775.mm.1 |
TC1500000862.mm.1 |
MTA TC |