Assay Details
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030225.4 | NP_084501.1 | 13 - 14 | 1127 | 133 | |
| GenBank mRNA | AK005477.1 | - | 1 - 2 | 104 | 133 | |
| AK019713.2 | - | 1124 | 133 | |||
| AK054053.1 | - | 13 - 14 | 1081 | 133 | ||
| AK149664.1 | - | 13 - 14 | 1103 | 133 | ||
| AK158877.1 | - | 13 - 14 | 1128 | 133 | ||
| AK168570.1 | - | 13 - 14 | 1103 | 133 | ||
| AK169943.1 | - | 13 - 14 | 1081 | 133 | ||
| BC006702.1 | - | 13 - 14 | 1064 | 3583936 | 133 | |
| BC024066.1 | - | 4 - 5 | 514 | 133 | ||
| CT010197.1 | - | 13 - 14 | 1063 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlst
Genomic Map