Assay Details
Gene Symbol: Ugt2b37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053215.3 | NP_444445.2 | 5 - 6 | 1329 | 159 | |
GenBank mRNA | AK002736.1 | - | 5 - 6 | 1346 | 159 | |
BC019487.1 | - | 5 - 6 | 1329 | 4237718 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302856 | NM_053215.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302856 | AK002736.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302856 | NM_053215.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302856 | AK002736.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539540 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39289 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632666 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589202 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853925 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15630 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76406 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171812 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302856 | NM_053215.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302856 | AK002736.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302856 | NM_053215.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302856 | AK002736.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23867086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23867098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23867112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23867089 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23867092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23867116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23867108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23867101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23867118 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05644445 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05644472 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05644463 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171812 | - | - |
Whole Mouse Genome G4122A | A_51_P102438 | BC145555.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102438 | NM_001029867.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102438 | AK149425.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102438 | BC141232.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102438 | BC145555.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102438 | NM_001029867.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102438 | AK149425.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102438 | BC141232.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302856 | NM_053215.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302856 | AK002736.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302856 | NM_053215.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302856 | AK002736.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112754 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131389 | BC069923.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131389 | AK034801.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131389 | BC015272.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131389 | BC057169.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131389 | NM_133894.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171812 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04176779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23867000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867110 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04879870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867094 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04618912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867116 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04126043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867087 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04318329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04631160 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867089 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04625207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867068 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867098 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04300571 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867112 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04879871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23867108 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10531064 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531065 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531062 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531063 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449890_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10531061 | - | - |
Mouse Genome 430 2.0 Array | 1449890_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449890_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 112417_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 112417_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500023787.mm.1 | TC0500002694.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023787.mm.1 | NM_053215.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023787.mm.1 | NM_053215.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023785.mm.1 | TC0500002694.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023785.mm.1 | NM_053215.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023785.mm.1 | NM_053215.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023788.mm.1 | TC0500002694.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023788.mm.1 | NM_053215.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023788.mm.1 | NM_053215.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023789.mm.1 | TC0500002694.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023789.mm.1 | NM_053215.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023789.mm.1 | NM_053215.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023784.mm.1 | TC0500002694.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023784.mm.1 | NM_053215.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023784.mm.1 | NM_053215.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023786.mm.1 | TC0500002694.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023786.mm.1 | NM_053215.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023786.mm.1 | NM_053215.3 | Refseq |
Mouse Expression Set 430 | 1449890_at | - | - |
Mouse Expression Set 430 | 1449890_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449890_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449890_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449890_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449890_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 927733 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4810204 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4810204 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004810204 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2729134 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2729134 | - | - |
mouse-6 v1.1 (Illumina) | scl0112417.1_237-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4810204 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0112417.1_237-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810204 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2729134 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2729134 | - | - |
