Assay Details
| Gene Symbol: Adamtsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301705.1 | NP_001288634.1 | 12 - 13 | 2369 | 72 | |
| NM_144899.3 | NP_659148.2 | 12 - 13 | 2251 | 72 | ||
| GenBank mRNA | AY158701.1 | - | 12 - 13 | 2192 | 72 | |
| BC117925.1 | - | 11 - 12 | 2164 | 40097608 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamtsl4
Genomic Map