Assay Details
| Gene Symbol: Adamtsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301705.1 | NP_001288634.1 | 13 - 14 | 2566 | 74 | |
| NM_144899.3 | NP_659148.2 | 13 - 14 | 2448 | 74 | ||
| GenBank mRNA | AK155481.1 | - | 13 - 14 | 2499 | 74 | |
| AY158701.1 | - | 13 - 14 | 2389 | 74 | ||
| BC117925.1 | - | 12 - 13 | 2361 | 40097608 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamtsl4
Genomic Map