Assay Details
Gene Symbol: Mustn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181390.3 | NP_852055.1 | 2 - 3 | 496 | 148 | |
GenBank mRNA | AJ277212.1 | - | 2 - 3 | 632 | 148 | |
AK003360.1 | - | 2 - 3 | 212 | 148 | ||
AK003945.1 | - | 2 - 3 | 214 | 148 | ||
BC116939.1 | - | 2 - 3 | 204 | 40126258 | 148 | |
BC116941.1 | - | 2 - 3 | 204 | 40126260 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | BC116939.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | AK003360.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | AK003945.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | BC116941.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | NM_181390.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | BC116939.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | AK003360.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | AK003945.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | BC116941.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P335064 | NM_181390.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967266 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118773 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | BC116939.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | AK003360.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | AK003945.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | BC116941.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | NM_181390.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | BC116939.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | AK003360.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | AK003945.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | BC116941.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P335064 | NM_181390.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508425 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508413 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508420 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508445 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508408 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508417 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508409 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508422 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508411 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508435 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508442 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508419 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29508438 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118773 | - | - |
Whole Mouse Genome G4122A | A_52_P335064 | BC116939.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335064 | AK003360.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335064 | AK003945.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335064 | BC116941.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335064 | NM_181390.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335064 | BC116939.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335064 | AK003360.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335064 | AK003945.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335064 | BC116941.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P335064 | NM_181390.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129881 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118773 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11592581 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11592557 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11592575 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11592572 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11592574 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11592545 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11592549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29508431 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508425 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11592581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11798386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508435 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508442 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508422 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508417 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11592586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508436 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508413 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11592574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508411 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508431 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508440 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11699526 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29508406 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10413613 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10413610 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10413614 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10413612 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10413611 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427201_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10413609 | - | - |
Mouse Genome 430 2.0 Array | 1427201_at | - | - |
Mouse Genome 430 2.0 Array | 1427201_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66175_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66175_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400002604.mm.1 | NM_181390.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400002604.mm.1 | NM_181390.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400002604.mm.1 | TC1400000316.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400002603.mm.1 | NM_181390.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400002603.mm.1 | NM_181390.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400002603.mm.1 | TC1400000316.mm.1 | MTA TC |
Mouse Expression Set 430 | 1427201_at | - | - |
Mouse Expression Set 430 | 1427201_PM_at | - | - |
Murine Genome U74v2 Set | 165443_f_at | - | - |
Murine Genome U74v2 Set | 165443_PM_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1427201_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427201_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427201_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427201_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 367701 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6380309 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006380309 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2658461 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2658461 | - | - |
mouse-6 v1.1 (Illumina) | scl46503.3.1_8-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6380309 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl46503.3.1_8-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6380309 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2658461 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2797928 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2658461 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2797928 | - | - |
