Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 1 - 2 | 440 | 114 | |
| NM_198438.1 | NP_940840.1 | 1 - 2 | 440 | 114 | ||
| GenBank mRNA | AF170906.1 | - | 1 - 2 | 170 | 114 | |
| AF500117.1 | - | 1 - 2 | 170 | 114 | ||
| AK011853.1 | - | 1 - 2 | 410 | 114 | ||
| AK017711.1 | - | 1 - 2 | 441 | 114 | ||
| BC003430.1 | - | 1 - 2 | 194 | 2646149 | 114 | |
| BC035475.1 | - | 1 - 2 | 383 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF170906.1, AF500117.1, AK011853.1, BC003430.1, NM_023672.2, NM_198438.1 transcripts.
Set Membership: |
|

Ssbp3
Genomic Map