Assay Details
| Gene Symbol: Apbb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253885.1 | NP_001240814.1 | 2 - 3 | 1089 | 74 | |
| NM_001253886.1 | NP_001240815.1 | 2 - 3 | 1089 | 74 | ||
| NM_009685.3 | NP_033815.1 | 2 - 3 | 903 | 74 | ||
| GenBank mRNA | AF206720.1 | - | 2 - 3 | 812 | 74 | |
| AK030748.1 | - | 2 - 3 | 786 | 74 | ||
| AK083830.1 | - | 2 - 3 | 819 | 74 | ||
| AK144340.1 | - | 2 - 3 | 1091 | 74 | ||
| AK164140.1 | - | 2 - 3 | 814 | 74 | ||
| BC036956.1 | - | 1 - 2 | 727 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apbb1
Genomic Map