Assay Details
| Gene Symbol: Apbb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253885.1 | NP_001240814.1 | 3 - 4 | 1267 | 127 | |
| NM_001253886.1 | NP_001240815.1 | 3 - 4 | 1267 | 127 | ||
| NM_001253887.1 | NP_001240816.1 | 3 - 4 | 267 | 127 | ||
| NM_009685.3 | NP_033815.1 | 3 - 4 | 1081 | 127 | ||
| GenBank mRNA | AF206720.1 | - | 3 - 4 | 990 | 127 | |
| AK030748.1 | - | 3 - 4 | 964 | 127 | ||
| AK075730.1 | - | 3 - 4 | 247 | 127 | ||
| AK083830.1 | - | 3 - 4 | 997 | 127 | ||
| AK144340.1 | - | 3 - 4 | 1268 | 126 | ||
| AK164140.1 | - | 3 - 4 | 992 | 127 | ||
| BC036956.1 | - | 2 - 3 | 905 | 127 | ||
| BC048395.1 | - | 2 - 3 | 325 | 6392016 | 127 | |
| BY240500.1 | - | 2 - 3 | 254 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC048395.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Apbb1
Genomic Map