Assay Details
| Gene Symbol: Mob4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025283.3 | NP_079559.2 | 2 - 3 | 177 | 103 | |
| GenBank mRNA | AK020446.1 | - | 2 - 3 | 133 | 103 | |
| AK147857.1 | - | 2 - 3 | 136 | 103 | ||
| AK162327.1 | - | 2 - 3 | 164 | 103 | ||
| BC028500.1 | - | 2 - 3 | 130 | 103 | ||
| BC037499.1 | - | 2 - 3 | 130 | 1512578 | 103 | |
| BC052475.1 | - | 2 - 3 | 122 | 103 | ||
| BC058168.1 | - | 2 - 3 | 130 | 4980300 | 103 | |
| BY768556.1 | - | 2 - 3 | 177 | 103 | ||
| U01138.1 | - | 2 - 3 | 121 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mob4
Genomic Map