Assay Details
| Gene Symbol: Ppp2r5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198168.3 | NP_937811.1 | 12 - 13 | 1844 | 71 | |
| GenBank mRNA | AK034167.1 | - | 12 - 13 | 1863 | 71 | |
| AK155872.1 | - | 12 - 13 | 1837 | 71 | ||
| AK171041.1 | - | 12 - 13 | 1815 | 71 | ||
| AK171178.1 | - | 12 - 13 | 1845 | 71 | ||
| AK171430.1 | - | 12 - 13 | 1852 | 71 | ||
| BC026670.1 | - | 8 - 9 | 700 | 71 | ||
| BC058977.1 | - | 12 - 13 | 1635 | 5687054 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r5b
Genomic Map